2-Thia-6-azaspiro[3.3]heptane-2,2-dioxide ≥95%

Supplier: Matrix Scientific

070679-250MG 070679-500MG 070679-1G
101945-870EA 1095.38 USD
101945-870 101945-872 101945-874
2-Thia-6-azaspiro[3.3]heptane-2,2-dioxide ≥95%
2-Thia-6-azaspiro[3.3]heptane-2,2-dioxide

Hazard: Irritant

Formula: C₅H₉NO₂S
MDL Number: MFCD17017544
CAS Number: 1263182-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR