1,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 1,6-diazaspiro[3.4]octane-6-carboxylate
070550-100MG
070550-250MG
070550-500MG
101945-262EA
367.69
USD
101945-262
101945-264
101945-266
1,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester ≥95%
1,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₁H₂₀N₂O₂ |
MDL Number:
MFCD12406509 CAS Number: 1158749-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...