Tributyl[2,2-difluoro-1-(2-methoxyethoxymethoxy)vinyl]stannane ≥97%

Supplier: Matrix Scientific

070527-1G 070527-5G
101945-164EA 1396.92 USD
101945-164 101945-166
Tributyl[2,2-difluoro-1-(2-methoxyethoxymethoxy)vinyl]stannane ≥97%
Tributyl[2,2-difluoro-1-(2-methoxyethoxymethoxy)vinyl]stannane

Hazard: Irritant

Formula: C₁₈H₃₆F₂O₃Sn
MDL Number: MFCD18325146
CAS Number: 170941-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR