4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

070247-1G 070247-5G
101943-944EA 1687.69 USD
101943-944 101943-946
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid ≥95%
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₇H₃ClN₂O₂S
MDL Number: MFCD11846188
CAS Number: 875515-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR