5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6,7,8-Tetrahydro-1,6-naphthyridine hcl

070244-1G 070244-5G
101943-932EA 1512.31 USD
101943-932 101943-934
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride ≥95%
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride

Hazard: Irritant

Formula: C₈H₁₁ClN₂
MDL Number: MFCD11846187
CAS Number: 1187830-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR