4-(4-Morpholinylcarbonyl)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid

070054-1G
101943-110EA 227.69 USD
101943-110
4-(4-Morpholinylcarbonyl)phenylboronic acid ≥98%
4-(4-Morpholinylcarbonyl)phenylboronic acid

Hazard: Irritant

Formula: C₁₁H₁₄BNO₄
Melting Pt: 126…132 °C
MDL Number: MFCD03411952
CAS Number: 389621-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR