3-(Hydroxymethyl)indole ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol

070050-5G
101943-096EA 73.85 USD
101943-096
3-(Hydroxymethyl)indole ≥98%
3-(Hydroxymethyl)indole

Hazard code(s): Irritant

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00005632
CAS Number: 700-06-1
EINECS: 211-836-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR