tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate ≥95%

Supplier: Matrix Scientific

069967-5G
101942-740EA 5163.08 USD
101942-740
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate ≥95%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₂S
MDL Number: MFCD08448157
CAS Number: 365996-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR