5-Bromoisoquinoline ≥98%

Supplier: Matrix Scientific
Warning

069778-5G
101941-890EA 38.46 USD
101941-890
5-Bromoisoquinoline ≥98%
5-Bromoisoquinoline

Hazard code(s): Irritant

Formula: C₉H₆BrN
MDL Number: MFCD01646405
CAS Number: 34784-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR