5-Bromo-1H-indazole ≥98%

Supplier: Matrix Scientific

069777-5G
101941-884EA 364.62 USD
101941-884
5-Bromo-1H-indazole ≥98%
5-Bromo-1H-indazole

Hazard code(s): Irritant

Formula: C₇H₅BrN₂
MDL Number: MFCD00839493
CAS Number: 53857-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR