Bis(triphenylphosphine)palladium(II) chloride ≥98%

Supplier: Matrix Scientific

Synonyms: Bis(triphenylphosphine)palladium(II) dichloride

069697-25G
101941-498EA 915.38 USD
101941-498
Bis(triphenylphosphine)palladium(II) chloride ≥98%
Bis(triphenylphosphine)palladium (II) chloride

Hazard code(s): Irritant, Hygroscopic, Refrigerate

Formula: C₃₆H₃₀Cl₂P₂Pd
MW: 701.91 g/mol
MDL Number: MFCD00009593
CAS Number: 13965-03-2
EINECS: 237-744-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR