3,4-Dichlorocyclobutene-1,2-dione ≥90%

Supplier: Matrix Scientific

Synonyms: 3,4-Dichlorocyclobut-3-ene-1,2-dione

069638-1G 069638-5G
101941-210EA 607.69 USD
101941-210 101941-212
3,4-Dichlorocyclobutene-1,2-dione ≥90%
3,4-Dichlorocyclobutene-1,2-dione

Hazard: Irritant

Formula: C₄Cl₂O₂
MDL Number: MFCD09763706
CAS Number: 2892-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR