4-Bromo-2-methoxythiazole ≥98%

Supplier: Matrix Scientific

069475-1G 069475-5G 069475-25G
101940-336EA 466.15 USD
101940-336 101940-338 101940-340
4-Bromo-2-methoxythiazole ≥98%
4-Bromo-2-methoxythiazole

Hazard: Irritant

Formula: C₄H₄BrNOS
MDL Number: MFCD07787374
CAS Number: 240816-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR