Murphy Plaster Knife, OR Grade, Sklar
Supplier: Sklar
11-1540
10194-972EA 73.63 USD
10194-972
Murphy Plaster Knife, OR Grade, Sklar
Knives
Murphy Plaster Knife features a wide handle and a wide, slightly curved blade used for removing pieces of a dried plaster cast
  • Made up of stainless steel
Censitrac Ready products are industry standard.
Instrumentation used for casting is typically stronger and heavier than standard instruments in order to adequately handle the heavy cast material. This plaster knife is equipped with a wooden handle.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.