Heinlein Expressor, OR Grade, Sklar
Supplier: Sklar
06-4186
10194-944EA 80.82 USD
10194-944
Heinlein Expressor, OR Grade, Sklar
Dissection Needles
This is a dermal instrument that features a lancet end for opening closed comedones and one spoon-shaped end with a hole for expressing comedones.
  • Reusable
  • Latex-free