Sklar Blue™ Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
91-5422
10194-322EA 400.42 USD
10194-322
Sklar Blue™ Dressing Forceps, OR Grade, Sklar
Forceps
Sklar Blue™ Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds during electrosurgical procedures.
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may be cleaned ultrasonically.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects. This product is straight with serrated tips and a length of 5 1/2".