Sklarlite XD™ Adson Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
23-2604
10194-222EA 112.94 USD
10194-222
Sklarlite XD™ Adson Dressing Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds
They have a wide thumb grasp for increased precision and control. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with serrated tips and a length of 4-3/4 inches