Micro-Cut Universal Style Biopsy Punch Kevorkian, OR Grade, Sklar
Supplier: Sklar
90-8966
10194-170EA 1899.61 USD
10194-170
Micro-Cut Universal Style Biopsy Punch Kevorkian, OR Grade, Sklar
Biopsy Punches
Sklar® Micro-Cut Universal Style Biopsy Punch is a reusable, stainless steel, premium OR grade punch.
  • Used in general surgery
  • Obstetrics and gynecology
This product is straight, has sharp tip, fenestrated edge style and pliers with spring handle style. This product measures 13.5” in length.