(3R,4R)-4-Amino-1-[[4-[(3-methoxyphenyl)amino]pyrrolo-[2,1-f][1,2,4]triazin-5-yl]methyl]piperidin-3-ol ≥95%

Supplier: Matrix Scientific

069274-100MG 069274-250MG 069274-500MG
101939-560EA 2850.77 USD
101939-560 101939-562 101939-564
(3R,4R)-4-Amino-1-[[4-[(3-methoxyphenyl)amino]pyrrolo-[2,1-f][1,2,4]triazin-5-yl]methyl]piperidin-3-ol ≥95%
(3R,4R)-4-Amino-1-[[4-[(3-methoxyphenyl)amino]pyrrolo-[2,1-f][1,2,4]triazin-5-yl]methyl]piperidin-3-ol

Hazard: Irritant

Formula: C₁₉H₂₄N₆O₂
MDL Number: MFCD18633295
CAS Number: 859853-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR