Cyclopamine ≥99%

Supplier: Matrix Scientific

Synonyms: (2'R,3S,3'R,3a'S,6aS,6bS,6'S,7a'R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3a',4,4',5',6,6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol

069242-25MG 069242-50MG 069242-100MG
101939-434EA 276.92 USD
101939-434 101939-436 101939-438
Cyclopamine ≥99%
Cyclopamine

Hazard: Irritant

Formula: C₂₇H₄₁NO₂
MDL Number: MFCD09878269
CAS Number: 4449-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR