5-Bromoquinoline ≥98% for synthesis

Supplier: Matrix Scientific
Warning

069144-5G 069144-25G
101939-074EA 610.77 USD
101939-074 101939-076
5-Bromoquinoline ≥98% for synthesis
5-Bromoquinoline

Hazard code(s): Irritant

Formula: C₉H₆BrN
MDL Number: MFCD00234481
CAS Number: 4964-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR