4-(4-Methoxyquinolin-2-yl)benzene-1,2-diol ≥95%

Supplier: Matrix Scientific

069117-250MG 069117-500MG
101938-998EA 1600 USD
101938-998 101939-000
4-(4-Methoxyquinolin-2-yl)benzene-1,2-diol ≥95%
4-(4-Methoxyquinolin-2-yl)benzene-1,2-diol

Hazard: Irritant

Formula: C₁₆H₁₃NO₃
MDL Number: MFCD19443960
CAS Number: 1313738-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR