tert-Butyl-10-benzyl-11-oxo-4,10-diazaspiro[5.5]undecane-4-carboxylate ≥95%

Supplier: Matrix Scientific

069018-250MG 069018-500MG 069018-1G
101938-648EA 698.46 USD
101938-648 101938-650 101938-652
tert-Butyl-10-benzyl-11-oxo-4,10-diazaspiro[5.5]undecane-4-carboxylate ≥95%
tert-Butyl-10-benzyl-11-oxo-4,10-diazaspiro[5.5]undecane-4-carboxylate

Hazard: Irritant

Formula: C₂₁H₃₀N₂O₃
MDL Number: MFCD12912610
CAS Number: 1198286-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR