3-Aminoisoxazole ≥95%

Supplier: Matrix Scientific

068857-25G
101937-968EA 670.77 USD
101937-968
3-Aminoisoxazole ≥95%
3-Aminoisoxazole

Hazard code(s): Irritant

Formula: C₃H₄N₂O
MDL Number: MFCD00038814
CAS Number: 1750-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR