3-Chloro-5-methyl-4-hydroxybenzonitrile ≥95%

Supplier: Matrix Scientific

068382-1G 068382-5G
101935-888EA 153.85 USD
101935-888 101935-890
3-Chloro-5-methyl-4-hydroxybenzonitrile ≥95%
3-Chloro-5-methyl-4-hydroxybenzonitrile

Hazard: Irritant

Formula: C₈H₆ClNO
MDL Number: MFCD08689583
CAS Number: 173900-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR