2-Methyl-6-pyrrolidin-2-ylpyridin-4-ol dihydrochloride

Supplier: Matrix Scientific

068175-250MG
101935-468EA 476.92 USD
101935-468
2-Methyl-6-pyrrolidin-2-ylpyridin-4-ol dihydrochloride
2-Methyl-6-pyrrolidin-2-ylpyridin-4-ol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂O
MDL Number: MFCD21606133
CAS Number: 1361116-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR