4-Methylpiperidine-4-carboxylic acid cyclopentylamide hydrochloride

Supplier: Matrix Scientific

067707-250MG
101934-532EA 476.92 USD
101934-532
4-Methylpiperidine-4-carboxylic acid cyclopentylamide hydrochloride
4-Methylpiperidine-4-carboxylic acid cyclopentylamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MDL Number: MFCD21605966
CAS Number: 1361116-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR