Methyl-4-fluoro-5-methoxy-2-nitrobenzoate

Supplier: Matrix Scientific

Synonyms: Methyl 4-fluoro-5-methoxy-2-nitrobenzoate

067200-500MG 067200-1G
101933-498EA 403.08 USD
101933-498 101933-500
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate

Hazard: Irritant

Formula: C₉H₈FNO₅
Melting Pt: 45…47 °C
MDL Number: MFCD20921594
CAS Number: 159768-50-0

Order Now

Specification Test Results

Melting point 45-47°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR