1-[2-(Thiomorpholin-4-yl)ethyl]imidazolidin-2-one ≥95%

Supplier: Matrix Scientific

066897-500MG 066897-1G
101932-682EA 600 USD
101932-682 101932-684
1-[2-(Thiomorpholin-4-yl)ethyl]imidazolidin-2-one ≥95%
1-[2-(Thiomorpholin-4-yl)ethyl]imidazolidin-2-one

Hazard: Irritant

Formula: C₉H₁₇N₃OS
Melting Pt: 129…131 °C
MDL Number: MFCD19982779
CAS Number: 1331823-96-1

Order Now

Specification Test Results

Melting point 129-131°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR