Ethyl-4-(chloromethyl)-1,3-thiazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

066797-500MG 066797-1G
101932-386EA 261.54 USD
101932-386 101932-388
Ethyl-4-(chloromethyl)-1,3-thiazole-2-carboxylate ≥95%
Ethyl-4-(chloromethyl)-1,3-thiazole-2-carboxylate

Hazard: Irritant

Formula: C₇H₈ClNO₂S
Melting Pt: 67…69 °C
MDL Number: MFCD10697515
CAS Number: 100960-16-5

Order Now

Specification Test Results

Melting point 67-69°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR