Benzyl-4-{[(3-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

066784-500MG 066784-1G
101932-334EA 600 USD
101932-334 101932-336
Benzyl-4-{[(3-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Benzyl-4-{[(3-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₃FN₂O₃
MDL Number: MFCD19686118
CAS Number: 1047655-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR