2-(4-Acetylpiperazin-1-yl)ethanamine

Supplier: Matrix Scientific

066688-1G
101932-140EA 638.46 USD
101932-140
2-(4-Acetylpiperazin-1-yl)ethanamine
2-(4-Acetylpiperazin-1-yl)ethanamine

Hazard: Irritant

Formula: C₈H₁₇N₃O
MDL Number: MFCD08275715
CAS Number: 148716-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR