1,2,3,4,5-Pentamethylcyclopentadiene ≥90%

Supplier: Matrix Scientific

066662-1G
101932-086EA 55.38 USD
101932-086
1,2,3,4,5-Pentamethylcyclopentadiene ≥90%
1,2,3,4,5-Pentamethylcyclopentadiene

Hazard code(s): Irritant

Formula: C₁₀H₁₆
Boiling Pt: 58 °C (44 hPa)
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
EINECS: 223-743-4
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR