2-(4-Pyridyl)benzimidazole

Supplier: Matrix Scientific

Synonyms: 2-(Pyridin-4-yl)-1H-benzo[d]imidazole

066595-500MG
101931-968EA 364.62 USD
101931-968
2-(4-Pyridyl)benzimidazole
2-(4-Pyridyl)benzimidazole

Hazard: Irritant

Formula: C₁₂H₉N₃
MDL Number: MFCD00453948
CAS Number: 2208-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR