3-(3,4-Dimethoxyphenyl)-7-hydroxy-4H-chromen-4-one

Supplier: Matrix Scientific

066517-500MG
101931-812EA 336.92 USD
101931-812
3-(3,4-Dimethoxyphenyl)-7-hydroxy-4H-chromen-4-one
3-(3,4-Dimethoxyphenyl)-7-hydroxy-4H-chromen-4-one

Hazard: Irritant

Formula: C₁₇H₁₄O₅
MDL Number: MFCD00076013
CAS Number: 24160-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR