N-(4-Bromophenyl)-N'-(4,6-dimethylpyrimidin-2-yl)guanidine

Supplier: Matrix Scientific

066427-500MG
101931-632EA 183.08 USD
101931-632
N-(4-Bromophenyl)-N'-(4,6-dimethylpyrimidin-2-yl)guanidine
N-(4-Bromophenyl)-N'-(4,6-dimethylpyrimidin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₃H₁₄BrN₅
MDL Number: MFCD01543005
CAS Number: 16018-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR