5-Ethyl-2-[(4-ethylphenyl)amino]-6-methylpyrimidin-4(3H)-one

Supplier: Matrix Scientific

066124-500MG
101931-028EA 364.62 USD
101931-028
5-Ethyl-2-[(4-ethylphenyl)amino]-6-methylpyrimidin-4(3H)-one
5-Ethyl-2-[(4-ethylphenyl)amino]-6-methylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₅H₁₉N₃O
MDL Number: MFCD19103568
CAS Number: 1306739-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR