2-Amino-6-(4-methylphenyl)pyrimidin-4(3H)-one

Supplier: Matrix Scientific

066111-500MG
101931-002EA 364.62 USD
101931-002
2-Amino-6-(4-methylphenyl)pyrimidin-4(3H)-one
2-Amino-6-(4-methylphenyl)pyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₁H₁₁N₃O
MW: 201.23 g/mol
MDL Number: MFCD13811750
CAS Number: 98305-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR