1-(2-Fluoro-4-methylphenyl)-1H-pyrrole-2,5-dione

Supplier: Matrix Scientific

066105-500MG
101930-990EA 243.08 USD
101930-990
1-(2-Fluoro-4-methylphenyl)-1H-pyrrole-2,5-dione
1-(2-Fluoro-4-methylphenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₁H₈FNO₂
MW: 205.19 g/mol
MDL Number: MFCD11184796
CAS Number: 1030420-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR