N-(2-Furylmethyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

066068-500MG
101930-916EA 364.62 USD
101930-916
N-(2-Furylmethyl)-1,3,5-triazine-2,4-diamine
N-(2-Furylmethyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₈H₉N₅O
MW: 191.19 g/mol
MDL Number: MFCD19103544
CAS Number: 98488-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR