3-(3-Chlorophenyl)quinazoline-2,4(1H,3H)-dione

Supplier: Matrix Scientific

065985-500MG
101930-750EA 243.08 USD
101930-750
3-(3-Chlorophenyl)quinazoline-2,4(1H,3H)-dione
3-(3-Chlorophenyl)quinazoline-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₄H₉ClN₂O₂
MDL Number: MFCD01859130
CAS Number: 2473-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR