4,4,6-Trimethyl-4,5-dihydro-1H-[1,2]dithiolo[3,4-c]quinoline-1-thione

Supplier: Matrix Scientific

065844-500MG
101930-468EA 243.08 USD
101930-468
4,4,6-Trimethyl-4,5-dihydro-1H-[1,2]dithiolo[3,4-c]quinoline-1-thione
4,4,6-Trimethyl-4,5-dihydro-1H-[1,2]dithiolo[3,4-c]quinoline-1-thione

Hazard: Irritant

Formula: C₁₃H₁₃NS₃
MW: 279.45 g/mol
MDL Number: MFCD00184282
CAS Number: 122246-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR