Gerzog Bone Mallet, OR Grade, Sklar
Supplier: Sklar
40-5765
10193-998EA 269.76 USD
10193-998
Gerzog Bone Mallet, OR Grade, Sklar
Mallets and Hammers
This lead filled Gerzog mallet is an orthopedic instrument used in conjuction with osteotomes, chisels and gouges to apply a swinging force when cutting or shaping bone
Censitrac Ready products are industry standard and constructed of stainless steel.
The Gerzog's face diameter is among the smaller versions, and it has a wide oval handle like the Kirk. This product is available individually, or as a component.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.