Pratt T-Shaped Clamp, OR Grade, Sklar®
Supplier: Sklar
17-1055
10193-926EA 346.15 USD
10193-926
Pratt T-Shaped Clamp, OR Grade, Sklar®
Clamps General Purpose Clamps
Pratt T-Shaped Clamp, also known as Pratt-Smith Grasping Forceps, is a ratcheted, finger ring, grasping instrument typically used in vaginal or perineal procedures for grasping mucosa or hemorrhoidal tissue
  • Constructed of stainless steel
Censitrac Ready products are industry standard.
The 16 cm concave tips of the clamp meet the jaws perpendicularly to create the t-shape that describes them. The serrated jaws provide a strong hold on tissues and may cause damage or crush tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.