Stitch Cutter, Sklar
Supplier: Sklar
06-3030
10193-782BX 121.93 USD
10193-782
Stitch Cutter, Sklar
Knives
The Stitch Cutter is used for general suture removal
Appearing similar to a scalpel, this instrument requires no handle and is a simple and convenient tool for removing basic sutures. The slightly curved, flat, bull nose tip slides under the suture and when rotated, gently removes the suture from the site. This product is the "midi" version and measures approximately 8.9cm (3.5") overall. Supplied sterile, in boxes of 100.