Laforce Foreign Body Spud, OR Grade, Sklar
Supplier: Sklar
65-7255
10193-384EA 162.64 USD
10193-384
Laforce Foreign Body Spud, OR Grade, Sklar
Dissection Needles
Sklar's Laforce Foreign Body Spud is an ophthalmic instrument typically used for removing foreign objects and debris from the eye.
This instrument is often referred to as the "golf club" spud because the shaft and tip resemble a golf club. This product has a flat, straight handle that meets a thin, straight shaft that tapers to a slightly angled 2 mm tip.