3-Methyl-4-(4-nitro-1H-pyrazol-1-yl)butan-2-one

Supplier: Matrix Scientific

065519-500MG
101929-838EA 484.62 USD
101929-838
3-Methyl-4-(4-nitro-1H-pyrazol-1-yl)butan-2-one
3-Methyl-4-(4-nitro-1H-pyrazol-1-yl)butan-2-one

Hazard: Irritant

Formula: C₈H₁₁N₃O₃
MDL Number: MFCD08457674
CAS Number: 942852-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR