3-Chloro-5-cyclohexyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

065477-500MG
101929-754EA 484.62 USD
101929-754
3-Chloro-5-cyclohexyl-1H-1,2,4-triazole
3-Chloro-5-cyclohexyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MW: 185.66 g/mol
MDL Number: MFCD18783023
CAS Number: 1279219-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR