N,N'-Dicyclohexylmorpholine-4-carboximidamide

Supplier: Matrix Scientific

065332-500MG
101929-464EA 243.08 USD
101929-464
N,N'-Dicyclohexylmorpholine-4-carboximidamide
N,N'-Dicyclohexylmorpholine-4-carboximidamide

Hazard: Irritant

Formula: C₁₇H₃₁N₃O
MW: 293.45 g/mol
MDL Number: MFCD00063252
CAS Number: 4975-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR