Docosanal ≥98%

Supplier: Matrix Scientific

064652-1G
101928-104EA 384.62 USD
101928-104
Docosanal ≥98%
Docosanal

Hazard: Irritant/Refrigerate

Formula: C₂₂H₄₄O
Melting Pt: 56…57 °C
MDL Number: MFCD07780595
CAS Number: 57402-36-5

Order Now

Specification Test Results

Melting point 56-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR